Assay Details
| Gene Symbol: TMEM11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018064825.1 | XP_017920314.1 | 2 | 502 | 101 | |
| GenBank mRNA | JO523256.2 | - | 1 | 443 | 101 | |
| KA318608.1 | - | 1 | 443 | 101 | ||
More Information
Set Membership: |
|

TMEM11
Genomic Map