Assay Details
| Gene Symbol: KCTD11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018064567.1 | XP_017920056.1 | 1 | 1109 | 91 | |
| GenBank mRNA | JT711550.1 | - | 5 | 1274 | 91 | |
| KA348710.1 | - | 1 | 1082 | 91 | ||
More Information
Set Membership: |
|

KCTD11
Genomic Map