Assay Details
| Gene Symbol: CD59 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | JO353893.2 | - | 4 | 1633 | 100 | |
| JO497236.2 | - | 213 | 100 | |||
| KA333945.1 | - | 1 | 194 | 100 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: CD59 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | JO353893.2 | - | 4 | 1633 | 100 | |
| JO497236.2 | - | 213 | 100 | |||
| KA333945.1 | - | 1 | 194 | 100 | ||
Set Membership: |
|