Assay Details
| Gene Symbol: LBX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018054993.1 | XP_017910482.1 | 1 | 224 | 125 | |
| GenBank mRNA | JT699837.1 | - | 1 | 140 | 125 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: LBX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018054993.1 | XP_017910482.1 | 1 | 224 | 125 | |
| GenBank mRNA | JT699837.1 | - | 1 | 140 | 125 | |
Set Membership: |
|