Assay Details
| Gene Symbol: BNIP3L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018052351.1 | XP_017907840.1 | 6 | 2625 | 99 | |
| XM_018052352.1 | XP_017907841.1 | 6 | 2600 | 99 | ||
| XM_018052353.1 | XP_017907842.1 | 6 | 2591 | 99 | ||
More Information
Set Membership: |
|

BNIP3L
Genomic Map