Assay Details
| Gene Symbol: SLC26A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018049876.1 | XP_017905365.1 | 3 | 5722 | 77 | |
| XM_018049877.1 | XP_017905366.1 | 3 | 5022 | 77 | ||
| XM_018049878.1 | XP_017905367.1 | 3 | 5682 | 77 | ||
| XM_018049879.1 | XP_017905368.1 | 4 | 3024 | 77 | ||
| GenBank mRNA | JO429418.2 | - | 1 | 1320 | 77 | |
More Information
Set Membership: |
|

SLC26A1
Genomic Map