Assay Details
| Gene Symbol: LOC108634231 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018043265.1 | XP_017898754.1 | 1 | 854 | 107 | |
| GenBank mRNA | JO539127.2 | - | 2 | 199 | 107 | |
| Gene Symbol: LOC102169782 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018045611.1 | XP_017901100.1 | 1 | 876 | 107 | |
| Gene Symbol: LOC102189503 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018060224.1 | XP_017915713.1 | 4 | 920 | 107 | |
| GenBank mRNA | JO562713.2 | - | 4 | 950 | 107 | |
More Information
Set Membership: |
|

LOC108634231
Genomic Map