Assay Details
| Gene Symbol: SST | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002758171.3 | XP_002758217.1 | 1 - 2 | 319 | 86 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SST| Gene Symbol: SST | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002758171.3 | XP_002758217.1 | 1 - 2 | 319 | 86 | |
Set Membership: |
|