Assay Details
| Gene Symbol: TRAF6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002755186.4 | XP_002755232.1 | 6 - 7 | 1035 | 64 | |
| XM_009007942.2 | XP_009006190.1 | 7 - 8 | 1246 | 64 | ||
| XM_009007943.2 | XP_009006191.1 | 7 - 8 | 1092 | 64 | ||
| XM_017978022.1 | XP_017833511.1 | 6 - 7 | 770 | 64 | ||
More Information
Set Membership: |
|

TRAF6
Genomic Map