Assay Details
| Gene Symbol: TBC1D20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002747417.3 | XP_002747463.1 | 2 - 3 | 397 | 79 | |
| XM_017971881.1 | XP_017827370.1 | 3 - 4 | 467 | 79 | ||
| XM_017971882.1 | XP_017827371.1 | 2 - 3 | 397 | 79 | ||
More Information
Set Membership: |
|

TBC1D20
Genomic Map