Assay Details
Gene Symbol: GRM7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002758591.3 | XP_002758637.1 | 1 - 2 | 884 | 72 | |
XM_003734881.3 | XP_003734929.1 | 1 - 2 | 883 | 72 | ||
XM_008982073.2 | XP_008980321.1 | 1 - 2 | 884 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
