Assay Details
| Gene Symbol: SH2D2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008984620.2 | XP_008982868.1 | 4 - 5 | 1019 | 55 | |
| XM_008984621.2 | XP_008982869.1 | 4 - 5 | 450 | 55 | ||
| XM_017966368.1 | XP_017821857.1 | 4 - 5 | 1025 | 55 | ||
More Information
Set Membership: |
|

SH2D2A
Genomic Map