Assay Details
Gene Symbol: TMEM237 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002749644.4 | XP_002749690.3 | 6 - 7 | 990 | 87 | |
XM_008999217.2 | XP_008997465.1 | 6 - 7 | 512 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() |
