Assay Details
| Gene Symbol: LOC100390922 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002757393.4 | XP_002757439.3 | 12 - 13 | 2043 | 76 | |
| XM_017964075.1 | XP_017819564.1 | 8 - 9 | 981 | 76 | ||
More Information
Set Membership: |
|

LOC100390922
Genomic Map