Assay Details
| Gene Symbol: SRPRB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003735067.3 | XP_003735115.1 | 4 - 5 | 642 | 68 | |
| XM_017965857.1 | XP_017821346.1 | 4 - 5 | 642 | 68 | ||
More Information
Set Membership: |
|

SRPRB
Genomic Map