Assay Details
| Gene Symbol: BRINP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002760271.4 | XP_002760317.1 | 3 - 4 | 908 | 112 | |
| XM_008984811.2 | XP_008983059.1 | 3 - 4 | 1908 | 112 | ||
| XM_008984813.2 | XP_008983061.1 | 3 - 4 | 1799 | 112 | ||
More Information
Set Membership: |
|

BRINP3
Genomic Map