Assay Details
| Gene Symbol: CNOT6L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008993110.2 | XP_008991358.1 | 5 - 6 | 639 | 67 | |
| XM_008993111.2 | XP_008991359.1 | 5 - 6 | 2818 | 67 | ||
| XM_017970471.1 | XP_017825960.1 | 5 - 6 | 634 | 67 | ||
More Information
Set Membership: |
|

CNOT6L
Genomic Map