Assay Details
| Gene Symbol: IGF2BP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017964733.1 | XP_017820222.1 | 1 - 2 | 308 | 97 | |
| XM_017964734.1 | XP_017820223.1 | 1 - 2 | 308 | 97 | ||
More Information
Set Membership: |
|

IGF2BP2
Genomic Map