Assay Details
| Gene Symbol: YWHAE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002747823.4 | XP_002747869.1 | 5 - 6 | 957 | 84 | |
| XM_008996386.1 | XP_008994634.1 | 6 - 7 | 891 | 84 | ||
More Information
Set Membership: |
|

YWHAE
Genomic Map