Assay Details
| Gene Symbol: MIF4GD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002748729.3 | XP_002748775.1 | 3 - 4 | 259 | 64 | |
| XM_008997739.1 | XP_008995987.1 | 3 - 4 | 506 | 64 | ||
| XM_008997740.2 | XP_008995988.1 | 3 - 4 | 415 | 64 | ||
| XM_008997741.2 | XP_008995989.1 | 3 - 4 | 421 | 64 | ||
| XM_008997742.1 | XP_008995990.1 | 3 - 4 | 320 | 64 | ||
| XM_017972832.1 | XP_017828321.1 | 3 - 4 | 276 | 64 | ||
More Information
Set Membership: |
|

MIF4GD
Genomic Map