Assay Details
| Gene Symbol: TRMT10B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008999713.2 | XP_008997961.1 | 4 - 5 | 540 | 85 | |
| XM_017974830.1 | XP_017830319.1 | 4 - 5 | 574 | 85 | ||
| XM_017974831.1 | XP_017830320.1 | 3 - 4 | 384 | 85 | ||
More Information
Set Membership: |
|

TRMT10B
Genomic Map