Assay Details
| Gene Symbol: ZNF207 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002748381.2 | XP_002748427.1 | 2 - 3 | 653 | 67 | |
| XM_002748382.2 | XP_002748428.1 | 2 - 3 | 653 | 67 | ||
| XM_002748383.3 | XP_002748429.1 | 2 - 3 | 653 | 67 | ||
| XM_008997108.1 | XP_008995356.1 | 2 - 3 | 653 | 67 | ||
| XM_008997109.1 | XP_008995357.1 | 2 - 3 | 653 | 67 | ||
| XM_017972538.1 | XP_017828027.1 | 2 - 3 | 653 | 67 | ||
More Information
Set Membership: |
|

ZNF207
Genomic Map