Assay Details
| Gene Symbol: TBXT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002747169.2 | XP_002747215.1 | 3 - 4 | 752 | 83 | |
| XM_002747170.3 | XP_002747216.1 | 3 - 4 | 607 | 83 | ||
More Information
Set Membership: |
|

TBXT
Genomic Map