Assay Details
| Gene Symbol: ZSWIM5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017974660.1 | XP_017830149.1 | 12 - 13 | 2675 | 61 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZSWIM5| Gene Symbol: ZSWIM5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017974660.1 | XP_017830149.1 | 12 - 13 | 2675 | 61 | |
Set Membership: |
|