Assay Details
| Gene Symbol: LOC100410987 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002756516.3 | XP_002756562.1 | 2 - 3 | 1285 | 99 | |
| XM_017979229.1 | XP_017834718.1 | 3 - 4 | 388 | 99 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017979229.1 transcripts.
Set Membership: |
|

LOC100410987
Genomic Map