Assay Details
| Gene Symbol: GALNT16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017977175.1 | XP_017832664.1 | 3 - 4 | 2142 | 64 | |
| XM_017977176.1 | XP_017832665.1 | 1 - 2 | 509 | 64 | ||
More Information
Set Membership: |
|

GALNT16
Genomic Map