Assay Details
Gene Symbol: GMEB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002750520.4 | XP_002750566.1 | 8 - 9 | 1856 | 76 | |
XM_003733387.3 | XP_003733435.1 | 8 - 9 | 1826 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
