Assay Details
| Gene Symbol: MBD5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008998924.2 | XP_008997172.1 | 5 - 6 | 1486 | 103 | |
| XM_017973489.1 | XP_017828978.1 | 2 - 3 | 784 | 103 | ||
| XM_017973490.1 | XP_017828979.1 | 6 - 7 | 1529 | 103 | ||
| XM_017973491.1 | XP_017828980.1 | 7 - 8 | 1165 | 103 | ||
| XM_017973492.1 | XP_017828981.1 | 6 - 7 | 1114 | 103 | ||
More Information
Set Membership: |
|

MBD5
Genomic Map