Assay Details
| Gene Symbol: DLAT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017977541.1 | XP_017833030.1 | 2 - 3 | 1019 | 103 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

DLAT| Gene Symbol: DLAT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017977541.1 | XP_017833030.1 | 2 - 3 | 1019 | 103 | |
Set Membership: |
|