Assay Details
| Gene Symbol: ZNF410 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002754139.4 | XP_002754185.2 | 8 - 9 | 1922 | 73 | |
| XM_002754140.4 | XP_002754186.1 | 8 - 9 | 1927 | 73 | ||
| XM_009006318.2 | XP_009004566.1 | 9 - 10 | 2076 | 73 | ||
| XM_017977205.1 | XP_017832694.1 | 7 - 8 | 1737 | 73 | ||
More Information
Set Membership: |
|

ZNF410
Genomic Map