Assay Details
| Gene Symbol: GAREM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002757156.3 | XP_002757202.1 | 4 - 5 | 1717 | 99 | |
| XM_008979663.2 | XP_008977911.1 | 4 - 5 | 1719 | 99 | ||
More Information
Set Membership: |
|

GAREM1
Genomic Map