Assay Details
| Gene Symbol: USP12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002748938.3 | XP_002748984.1 | 4 - 5 | 826 | 117 | |
| XM_008998055.2 | XP_008996303.1 | 7 - 8 | 998 | 117 | ||
| XM_017973008.1 | XP_017828497.1 | 8 - 9 | 927 | 117 | ||
| XM_017973009.1 | XP_017828498.1 | 7 - 8 | 766 | 117 | ||
| XM_017973010.1 | XP_017828499.1 | 7 - 8 | 861 | 117 | ||
More Information
Set Membership: |
|

USP12
Genomic Map