Assay Details
| Gene Symbol: CDC5L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002746594.4 | XP_002746640.1 | 7 - 8 | 1200 | 120 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CDC5L| Gene Symbol: CDC5L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002746594.4 | XP_002746640.1 | 7 - 8 | 1200 | 120 | |
Set Membership: |
|