Assay Details
| Gene Symbol: CDS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002747421.4 | XP_002747467.2 | 5 - 6 | 868 | 77 | |
| XM_008995717.2 | XP_008993965.2 | 5 - 6 | 2461 | 77 | ||
| XM_017971948.1 | XP_017827437.1 | 6 - 7 | 2258 | 77 | ||
More Information
Set Membership: |
|

CDS2
Genomic Map