Assay Details
| Gene Symbol: NFIA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002750888.2 | XP_002750934.1 | 5 - 6 | 1274 | 66 | |
| XM_002750889.3 | XP_002750935.1 | 6 - 7 | 1067 | 66 | ||
| XM_009001349.1 | XP_008999597.1 | 5 - 6 | 1230 | 66 | ||
| XM_009001350.1 | XP_008999598.1 | 5 - 6 | 1230 | 66 | ||
| XM_009001351.1 | XP_008999599.1 | 5 - 6 | 1230 | 66 | ||
| XM_017974797.1 | XP_017830286.1 | 5 - 6 | 920 | 66 | ||
More Information
Set Membership: |
|

NFIA
Genomic Map