Assay Details
| Gene Symbol: FPGT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001204801.2 | NP_001191730.2 | 2 - 3 | 321 | 86 | |
| XM_009001485.2 | XP_008999733.1 | 2 - 3 | 310 | 86 | ||
More Information
Set Membership: |
|

FPGT
Genomic Map