Assay Details
| Gene Symbol: ANGPTL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002760331.4 | XP_002760377.1 | 2 - 3 | 5531 | 80 | |
| XM_017966045.1 | XP_017821534.1 | 1 - 2 | 6034 | 80 | ||
More Information
Set Membership: |
|

ANGPTL1
Genomic Map