Assay Details
| Gene Symbol: NAPG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002757049.3 | XP_002757095.1 | 3 - 4 | 447 | 67 | |
| XM_017963860.1 | XP_017819349.1 | 3 - 4 | 167 | 67 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017963860.1 transcripts.
Set Membership: |
|

NAPG
Genomic Map