Assay Details
| Gene Symbol: RBM45 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002749274.3 | XP_002749320.2 | 1 - 2 | 412 | 58 | |
| XM_008998640.1 | XP_008996888.1 | 1 - 2 | 412 | 58 | ||
| XM_008998641.1 | XP_008996889.1 | 1 - 2 | 412 | 58 | ||
| XM_008998642.1 | XP_008996890.1 | 1 - 2 | 412 | 58 | ||
| XM_008998643.1 | XP_008996891.1 | 1 - 2 | 412 | 58 | ||
| XM_008998644.1 | XP_008996892.1 | 1 - 2 | 412 | 58 | ||
| XM_008998645.1 | XP_008996893.1 | 1 - 2 | 412 | 58 | ||
| XM_008998646.1 | XP_008996894.1 | 1 - 2 | 412 | 58 | ||
| XM_008998647.1 | XP_008996895.1 | 1 - 2 | 412 | 58 | ||
More Information
Set Membership: |
|

RBM45
Genomic Map