Assay Details
| Gene Symbol: CAMK1G | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002760756.2 | XP_002760802.1 | 3 - 4 | 351 | 64 | |
| XM_008985611.2 | XP_008983859.1 | 3 - 4 | 403 | 64 | ||
| XM_008985612.2 | XP_008983860.1 | 3 - 4 | 375 | 64 | ||
| XM_008985613.2 | XP_008983861.1 | 3 - 4 | 1547 | 64 | ||
More Information
Set Membership: |
|

CAMK1G
Genomic Map