Assay Details
| Gene Symbol: SLC9B1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017970384.1 | XP_017825873.1 | 3 - 4 | 785 | 95 | |
| XM_017970385.1 | XP_017825874.1 | 9 - 10 | 4851 | 95 | ||
More Information
Set Membership: |
|

SLC9B1
Genomic Map