Assay Details
| Gene Symbol: AK3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002742903.4 | XP_002742949.1 | 1 - 2 | 374 | 62 | |
| XM_008996514.2 | XP_008994762.1 | 1 - 2 | 374 | 62 | ||
More Information
Set Membership: |
|

AK3
Genomic Map