Assay Details
| Gene Symbol: BRMS1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_009005939.2 | XP_009004187.1 | 8 - 9 | 918 | 103 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

BRMS1L| Gene Symbol: BRMS1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_009005939.2 | XP_009004187.1 | 8 - 9 | 918 | 103 | |
Set Membership: |
|