Assay Details
| Gene Symbol: WASHC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002752907.2 | XP_002752953.1 | 6 - 7 | 664 | 92 | |
| XM_009004489.1 | XP_009002737.1 | 6 - 7 | 661 | 92 | ||
More Information
Set Membership: |
|

WASHC3
Genomic Map