Assay Details
| Gene Symbol: LDHC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_009007797.2 | XP_009006045.1 | 5 - 6 | 755 | 67 | |
| XM_009007798.2 | XP_009006046.1 | 5 - 6 | 707 | 67 | ||
More Information
Set Membership: |
|

LDHC
Genomic Map