Assay Details
| Gene Symbol: LOC100406494 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017976341.1 | XP_017831830.1 | 1 - 2 | 2284 | 109 | |
| XM_017976342.1 | XP_017831831.1 | 1 - 2 | 2285 | 109 | ||
More Information
Set Membership: |
|

LOC100406494
Genomic Map