Assay Details
| Gene Symbol: PREB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002757944.4 | XP_002757990.1 | 4 - 5 | 877 | 64 | |
| XM_008980996.2 | XP_008979244.1 | 4 - 5 | 846 | 64 | ||
| XM_008980997.2 | XP_008979245.1 | 4 - 5 | 625 | 64 | ||
| XM_017964472.1 | XP_017819961.1 | 4 - 5 | 784 | 64 | ||
More Information
Set Membership: |
|

PREB
Genomic Map