Assay Details
| Gene Symbol: NABP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_009003984.2 | XP_009002232.1 | 2 - 3 | 210 | 70 | |
| XM_017976142.1 | XP_017831631.1 | 1 - 2 | 551 | 70 | ||
More Information
Set Membership: |
|

NABP2
Genomic Map