Assay Details
| Gene Symbol: B4GALT3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002760191.3 | XP_002760237.1 | 3 - 4 | 988 | 68 | |
| XM_002760192.4 | XP_002760238.2 | 3 - 4 | 967 | 68 | ||
| XM_008984772.2 | XP_008983020.1 | 3 - 4 | 1529 | 68 | ||
More Information
Set Membership: |
|

B4GALT3
Genomic Map