Assay Details
| Gene Symbol: WSCD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002752974.4 | XP_002753020.4 | 6 - 7 | 1970 | 72 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

WSCD2| Gene Symbol: WSCD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002752974.4 | XP_002753020.4 | 6 - 7 | 1970 | 72 | |
Set Membership: |
|